2,3-Difluoroaniline ≥97.0%

Supplier: TCI America

D2595-1G D2595-5G
TCD2595-001GEA 43 USD
TCD2595-001G TCD2595-005G
2,3-Difluoroaniline ≥97.0%
2,3-Difluoroaniline

CAS Number: 4519-40-8
MDL Number: MFCD00010298
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 71
Specific Gravity (20/20): 1.28

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 69 °C
Density: 1.28 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00010298
CAS Number: 4519-40-8
EINECS: 224-847-2
UN: 2941
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d14a6c75bdf81607c11365d3e7ef2eb025f8893&vid=934077a6db49650a445ee8516eb6148cc95b0468&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR