4-Nitro-2,6-xylenol ≥98.0%
Supplier: TCI America
CAS Number: 2423-71-4
MDL Number: MFCD00007339
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 170
Other Product Names / Synonyms: 4-Nitro-2,6-xylenol
Formula:
C₈H₉NO₃ Melting Pt: 170 °C |
MDL Number:
MFCD00007339 CAS Number: 2423-71-4 EINECS: 219-353-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...