2,4-Dichlorophenylhydrazinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2,4-Dichlorophenyl)hydrazine hydrochloride, 2,4-Dichlorophenylhydrazine hydrochloride, 1-(2,4-Dichlorophenyl)hydrazine hydrochloride

D2433-5G D2433-25G
TCD2433-005GEA 46 USD
TCD2433-005G TCD2433-025G
2,4-Dichlorophenylhydrazinium chloride ≥98.0% (by titrimetric analysis)
2,4-Dichlorophenylhydrazinium chloride

CAS Number: 5446-18-4
MDL Number: MFCD00012929
Molecular Formula: C6H6Cl2N2
Molecular Weight: 213.49
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
MDL Number: MFCD00012929
CAS Number: 5446-18-4
EINECS: 226-659-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=365304af6ec07b0dfb19699b9cddce6bdd7e0ebc&vid=a0d4d4439cc13c5fe4f7ca69de05f4daf6660259&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR