3,5-Di-tert-butyl-o-benzoquinone ≥98.0%
Supplier: TCI America
D2430-5G
D2430-25G
TCD2430-005GEA
90.54
USD
TCD2430-005G
TCD2430-025G
3,5-Di-tert-butyl-o-benzoquinone ≥98.0%
3,5-Di-tert-butyl-o-benzoquinone
CAS Number: 3383-21-9
MDL Number: MFCD00001647
Molecular Formula: C14H20O2
Molecular Weight: 220.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 115
Formula:
C₁₄H₂₀O₂ MW: 220.31 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00001647 CAS Number: 3383-21-9 EINECS: 222-189-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...