5,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5,6-Dichloro-3-pyridinecarboxylic acid, 5,6-Dichloropyridine-3-carboxylic acid, 5,6-Dichloropyridine-3-carboxylicacid

D2401-25G D2401-5G
TCD2401-25GEA 179.9 USD
TCD2401-25G TCD2401-005G
5,6-Dichloronicotinic acid ≥98.0% (by titrimetric analysis)
5,6-Dichloronicotinic acid

CAS Number: 41667-95-2
MDL Number: MFCD00075181
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 5,6-Dichloro-3-pyridinecarboxylic Acid

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00075181
CAS Number: 41667-95-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ca8a96d3751b831cef125dd37f943238649019&vid=25ca8a96d3751b831cef125dd37f943238649019&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR