6-Nitro-2,4-xylenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,4-Dimethyl-6-nitrophenol, 4-Hydroxy-5-nitro-3-xylene
D2273-25G
D2273-5G
TCD2273-25GEA
246.9
USD
TCD2273-25G
TCD2273-005G
6-Nitro-2,4-xylenol ≥98.0% (by GC, titration analysis)
6-Nitro-2,4-xylenol
CAS Number: 14452-34-7
MDL Number: MFCD00191633
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 71
Other Product Names / Synonyms: 6-Nitro-2,4-xylenol
Formula:
C₈H₉NO₃ MW: 167.16 g/mol Melting Pt: 71 °C |
MDL Number:
MFCD00191633 CAS Number: 14452-34-7 EINECS: 238-436-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...