Diphenyl diselenide ≥97.0%

Supplier: TCI America
Danger

D2186-5G D2186-25G
TCD2186-005GEA 36.8 USD
TCD2186-005G TCD2186-025G
Diphenyl diselenide ≥97.0%
Diphenyl diselenide

CAS Number: 1666-13-3
MDL Number: MFCD00003001
Molecular Formula: C12H10Se2
Molecular Weight: 312.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 202
Melting point (°C): 61


Product Categories: Synthetic Organic Chemistry, Selenenylation
Other Product Names / Synonyms: Phenyl Diselenide

Formula: C₁₂H₁₀Se₂
Boiling Pt: 202 °C
Melting Pt: 61 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003001
CAS Number: 1666-13-3
EINECS: 216-780-2
UN: 3283
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5548dcb38641f0922f8ef06d9c76890739673d2a&vid=5548dcb38641f0922f8ef06d9c76890739673d2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR