2,5-Dimethylpyrazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D2171-25ML D2171-500ML D2171-100ML
TCD2171-025MLEA 49 USD
TCD2171-025ML TCD2171-500ML TCD2171-100ML
2,5-Dimethylpyrazine ≥98.0% (by GC, titration analysis)
2,5-Dimethylpyrazine

CAS Number: 123-32-0
MDL Number: MFCD00006147
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 64
Specific Gravity (20/20): 0.99

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 155 °C
Melting Pt: 15 °C
Density: 0.99 (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c8c466f0f8007c07ffefde5e67d001e42d040ed&vid=2c8c466f0f8007c07ffefde5e67d001e42d040ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR