2,6-Dichlorobenzoyl chloride ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

D2064-500G D2064-25G
TCD2064-500GEA 416.4 USD
TCD2064-500G TCD2064-25G
2,6-Dichlorobenzoyl chloride ≥98.0%
2,6-Dichlorobenzoyl chloride

CAS Number: 4659-45-4
MDL Number: MFCD00000662
Molecular Formula: C7H3Cl3O
Molecular Weight: 209.45
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 147
Specific Gravity (20/20): 1.47

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 127 °C
Melting Pt: 18 °C
Density: 1.47 (20 °C)
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4feb89aaa3979701564b7d4b733941aec51a03ba&vid=4feb89aaa3979701564b7d4b733941aec51a03ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR