1,3-Dichloro-4-fluorobenzene ≥96.0%

Supplier: TCI America
Warning

D2059-500G D2059-25G
TCD2059-500GEA 223.9 USD
TCD2059-500G TCD2059-25G
1,3-Dichloro-4-fluorobenzene ≥96.0%
1,3-Dichloro-4-fluorobenzene

CAS Number: 1435-48-9
MDL Number: MFCD00000580
Molecular Formula: C6H3Cl2F
Molecular Weight: 164.99
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 37
Specific Gravity (20/20): 1.41

Formula: C₆H₃Cl₂F
Boiling Pt: 173 °C
Melting Pt: -23 °C
Density: 1.41 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00000580
CAS Number: 1435-48-9
EINECS: 406-160-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92e7fdd1f523b3560a6b57774adb1c42b3b4c9fb&vid=128a2bfe48c4d61ed0533f6b546bbe9135b327ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR