(±)-Ethyl-2,3-dibromopropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2,3-Dibromopropionate

D1992-500G D1992-25G
TCD1992-500GEA 165.3 USD
TCD1992-500G TCD1992-25G
(±)-Ethyl-2,3-dibromopropionate ≥98.0%
(±)-Ethyl-2,3-dibromopropionate

CAS Number: 3674-13-3
MDL Number: MFCD00000212
Molecular Formula: C5H8Br2O2
Molecular Weight: 259.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 91
Specific Gravity (20/20): 1.79


Other Product Names / Synonyms: 2,3-Dibromopropionic Acid Ethyl Ester

Formula: C₅H₈Br₂O₂
MW: 259.93 g/mol
Boiling Pt: 82 °C
Density: 1.79 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00000212
CAS Number: 3674-13-3
EINECS: 222-941-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=364b63c7dd5ca7f591c0c70f6a341d7d8ff8e788&vid=364b63c7dd5ca7f591c0c70f6a341d7d8ff8e788&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR