3-(3,4-Dimethoxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid

D1953-25G D1953-10G
TCD1953-25GEA 109 USD
TCD1953-25G TCD1953-010G
3-(3,4-Dimethoxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)
3-(3,4-Dimethoxyphenyl)propionic acid

CAS Number: 2107-70-2
MDL Number: MFCD00002774
Molecular Formula: C11H14O4
Molecular Weight: 210.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 99


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3,4-Dimethoxyhydrocinnamic Acid

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 99 °C
MDL Number: MFCD00002774
CAS Number: 2107-70-2
EINECS: 218-288-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8afd1d24d6090c32dedcc2980ce90396d1ef3312&vid=8afd1d24d6090c32dedcc2980ce90396d1ef3312&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR