3',4'-Dimethoxyacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone

D1878-500G D1878-25G D1878-100G
TCD1878-500GEA 376 USD
TCD1878-500G TCD1878-025G TCD1878-100G
3',4'-Dimethoxyacetophenone ≥98.0%
3',4'-Dimethoxyacetophenone

CAS Number: 1131-62-0
MDL Number: MFCD00008737
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 161
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Acetylveratrole

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 161 °C
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00008737
CAS Number: 1131-62-0
EINECS: 214-468-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f91fc112a2d1861343113f6d16d6d2d2ecc7f75&vid=8f91fc112a2d1861343113f6d16d6d2d2ecc7f75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR