3,4-Dichloro-α,α,α-trifluorotoluene ≥98.0%
Supplier: TCI America
|
Warning
|
D1865-25G
D1865-500G
TCD1865-025GEA
19
USD
TCD1865-025G
TCD1865-500G
3,4-Dichloro-α,α,α-trifluorotoluene ≥98.0%
3,4-Dichloro-α,α,α-trifluorotoluene
CAS Number: 328-84-7
MDL Number: MFCD00000555
Molecular Formula: C7H3Cl2F3
Molecular Weight: 215.00
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 65
Specific Gravity (20/20): 1.49
Formula:
C₇H₃Cl₂F₃ Boiling Pt: 173 °C Melting Pt: –0.2 °C Density: 1.49 (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000555 CAS Number: 328-84-7 EINECS: 206-337-1 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...