N,N-Dimethyloctylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
N,N-Dimethyloctan-1-amine
D1814-25ML
D1814-500ML
TCD1814-25MLEA
51.13
USD
TCD1814-25ML
TCD1814-500ML
N,N-Dimethyloctylamine ≥97.0% (by GC, titration analysis)
N,N-Dimethyloctylamine
CAS Number: 7378-99-6
MDL Number: MFCD00009558
Molecular Formula: C10H23N
Molecular Weight: 157.30
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 90
Melting point (°C): -57
Flash Point (°C): 64
Specific Gravity (20/20): 0.77
Other Product Names / Synonyms: N-n-Octyldimethylamine
Formula:
C₁₀H₂₃N MW: 157.3 g/mol Boiling Pt: 90 °C Melting Pt: -57 °C Density: 0.77 (20 °C) Flash Pt: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009558 CAS Number: 7378-99-6 EINECS: 230-939-3 UN: 2922 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...