1,3-Dichloro-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D1783-500G D1783-25G D1783-100G
TCD1783-500GEA 62 USD
TCD1783-500G TCD1783-25G TCD1783-100G
1,3-Dichloro-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)
1,3-Dichloro-5,5-dimethylhydantoin

CAS Number: 118-52-5
MDL Number: MFCD00003190
Molecular Formula: C5H6Cl2N2O2
Molecular Weight: 197.02
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 132

Formula: C₅H₆Cl₂N₂O₂
Melting Pt: 132 °C
MDL Number: MFCD00003190
CAS Number: 118-52-5
EINECS: 204-258-7
UN: 1479
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57939279b7873d1841ef7790173c4621385de127&vid=57939279b7873d1841ef7790173c4621385de127&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR