meso-2,3-Dimercaptosuccinic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1722-5G D1722-25G
TCD1722-005GEA 86 USD
TCD1722-005G TCD1722-025G
meso-2,3-Dimercaptosuccinic acid ≥98.0% (by titrimetric analysis)
meso-2,3-Dimercaptosuccinic acid

CAS Number: 304-55-2
MDL Number: MFCD00064799
Molecular Formula: C4H6O4S2
Molecular Weight: 182.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 198

Formula: C₄H₆O₄S₂
Melting Pt: 198 °C
Storage Temperature: Freezer
MDL Number: MFCD00064799
CAS Number: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2fa3535e105e5fb9dbb2d3d4e97441cec9a5a2c9&vid=2fa3535e105e5fb9dbb2d3d4e97441cec9a5a2c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR