3,5-Diiodosalicylic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-3,5-diiodobenzoic acid

D1677-250G D1677-25G
TCD1677-250GEA 199 USD
TCD1677-250G TCD1677-025G
3,5-Diiodosalicylic acid ≥98.0%
3,5-Diiodosalicylic acid

CAS Number: 133-91-5
MDL Number: MFCD00002444
Molecular Formula: C7H4I2O3
Molecular Weight: 389.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 230

Formula: C₇H₄I₂O₃
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00002444
CAS Number: 133-91-5
EINECS: 205-124-0
Merck Index: 13,03212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a79c27f77e8598e3bb444fcf0d09dda2462b615&vid=6238559d2afdcab1d42981b00ef841087a3ba6f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR