3,4-Dibromothiophene ≥98.0%
Supplier: TCI America
|
Warning
|
D1676-25G
D1676-5G
TCD1676-25GEA
155.9
USD
TCD1676-25G
TCD1676-005G
3,4-Dibromothiophene ≥98.0%
3,4-Dibromothiophene
CAS Number: 3141-26-2
MDL Number: MFCD00005465
Molecular Formula: C4H2Br2S
Molecular Weight: 241.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): 5
Flash Point (°C): 110
Specific Gravity (20/20): 2.21
Formula:
C₄H₂Br₂S MW: 241.93 g/mol Boiling Pt: 100 °C Melting Pt: 5 °C Density: 2.21 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005465 CAS Number: 3141-26-2 EINECS: 221-546-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...