2,6-Di-tert-butyl-4-ethylphenol ≥98.0%
Supplier: TCI America
|
Warning
|
D1667-500G
D1667-25G
D1667-100G
TCD1667-500GEA
302
USD
TCD1667-500G
TCD1667-25G
TCD1667-100G
2,6-Di-tert-butyl-4-ethylphenol ≥98.0%
2,6-Di-tert-butyl-4-ethylphenol
CAS Number: 4130-42-1
MDL Number: MFCD00017366
Molecular Formula: C16H26O
Molecular Weight: 234.38
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 137
Melting point (°C): 45
Formula:
C₁₆H₂₆O Boiling Pt: 137 °C Melting Pt: 45 °C |
MDL Number:
MFCD00017366 CAS Number: 4130-42-1 EINECS: 223-945-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...