Ethyl-1,3-dithiane-2-carboxylate ≥98.0%

Supplier: TCI America

D1648-1G D1648-5G
TCD1648-001GEA 85 USD
TCD1648-001G TCD1648-5G
Ethyl-1,3-dithiane-2-carboxylate ≥98.0%
Ethyl 1,3-dithiane-2-carboxylate

CAS Number: 20462-00-4
MDL Number: MFCD00006657
Molecular Formula: C7H12O2S2
Molecular Weight: 192.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 55
Specific Gravity (20/20): 1.22


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 1,3-Dithiane-2-carboxylic Acid Ethyl Ester

Formula: C₇H₁₂O₂S₂
Density: 1.22 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00006657
CAS Number: 20462-00-4
EINECS: 243-838-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06c89eb95e3ca3090935378478823d07e3cb5623&vid=06c89eb95e3ca3090935378478823d07e3cb5623&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR