1,3-Difluoro-4-nitrobenzene ≥99.0%

Supplier: TCI America

Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene

D1636-500G D1636-25G
TCD1636-500GEA 148 USD
TCD1636-500G TCD1636-025G
1,3-Difluoro-4-nitrobenzene ≥99.0%
1,3-Difluoro-4-nitrobenzene

CAS Number: 446-35-5
MDL Number: MFCD00007050
Molecular Formula: C6H3F2NO2
Molecular Weight: 159.09
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 90
Freezing point (°C): 9
Specific Gravity (20/20): 1.45

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 207 °C
Melting Pt: 10 °C
Density: 1.45 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00007050
CAS Number: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d44b302a084181b91cbfd64addf24ead58824f2&vid=42b76981eccc32460505687df0a33ba7ec795e13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR