2,6-Difluoroaniline ≥98.0%
Supplier: TCI America
|
Danger
|
D1635-5G
D1635-25G
TCD1635-005GEA
27
USD
TCD1635-005G
TCD1635-025G
2,6-Difluoroaniline ≥98.0%
2,6-Difluoroaniline
CAS Number: 5509-65-9
MDL Number: MFCD00007655
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 43
Specific Gravity (20/20): 1.28
Formula:
C₆H₅F₂N Boiling Pt: 154 °C Density: 1.28 (20 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007655 CAS Number: 5509-65-9 EINECS: 226-853-0 UN: 1992 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...