Di-n-Alkyldimethylammonium chloride (mixture) ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Di(hydrogenated tallow)dimethylammonium chloride, Quaternium-18, Arquad® 2HT-75, Dimethyldioctadecylammonium chloride, mixture of homologs

D1630-25G D1630-500G
TCD1630-025GEA 26 USD
TCD1630-025G TCD1630-500G
Di-n-Alkyldimethylammonium chloride (mixture) ≥95.0% (by titrimetric analysis)
Di-n-Alkyldimethylammonium chloride (mixture)

CAS Number: 61789-80-8 CAS Number: 61789-80-8 Form: Crystal

Formula: C₃₈H₈₀ClN
MDL Number: MFCD01775729
CAS Number: 107-64-2
EINECS: 263-090-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=629830bc9e86512b809b880534316b3a8d598958&vid=7bf2b255c00db8222ac51a6ad17aac3b3be97791&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR