1,3-Difluorobenzene ≥98.0%

Supplier: TCI America

D1626-500G D1626-25G
TCD1626-500GEA 458 USD
TCD1626-500G TCD1626-25G
1,3-Difluorobenzene ≥98.0%
1,3-Difluorobenzene

CAS Number: 372-18-9
MDL Number: MFCD00000327
Molecular Formula: C6H4F2
Molecular Weight: 114.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Melting point (°C): -59
Flash Point (°C): -5
Specific Gravity (20/20): 1.16

Formula: C₆H₄F₂
Boiling Pt: 83 °C
Melting Pt: –59 °C
Density: 1.16 (20 °C)
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00000327
CAS Number: 372-18-9
EINECS: 206-746-5
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58e3541790d72916886f9bcd12bdbde2361eb20&vid=b33809b8cb10b93bbbbf4d83376b2eb4fccd0061&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR