Bis(cyclopentadienyl)nickel ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Nickelocene

D1574-5G D1574-1G
TCD1574-005GEA 163 USD
TCD1574-005G TCD1574-001G
Bis(cyclopentadienyl)nickel ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)nickel

CAS Number: 1271-28-9
MDL Number: MFCD00001441
Molecular Formula: C10H10Ni
Molecular Weight: 188.88
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: Bis(cyclopentadienyl)nickel

Formula: C₁₀H₁₀Ni
MW: 188.88 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00001441
CAS Number: 1271-28-9
EINECS: 215-039-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6cac9b209ebc5874de834f8cc6228952b32467c&vid=d6cac9b209ebc5874de834f8cc6228952b32467c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR