1,2-Dimethylimidazole ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1,2-Dimethyl-1H-imidazole
D1562-500G
D1562-25G
D1562-100G
TCD1562-500GEA
207
USD
TCD1562-500G
TCD1562-25G
TCD1562-100G
1,2-Dimethylimidazole ≥98.0%
1,2-Dimethylimidazole
CAS Number: 1739-84-0
MDL Number: MFCD00005294
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 205
Freezing point (°C): 37
Formula:
C₅H₈N₂ MW: 96.13 g/mol Boiling Pt: 205 °C Melting Pt: 38 °C Density: 1.084 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005294 CAS Number: 1739-84-0 EINECS: 217-101-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...