2,4-Difluoroaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1508-25G D1508-500G
TCD1508-025GEA 50.88 USD
TCD1508-025G TCD1508-500G
2,4-Difluoroaniline ≥98.0% (by GC, titration analysis)
2,4-Difluoroaniline

CAS Number: 367-25-9
MDL Number: MFCD00007648
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 62
Specific Gravity (20/20): 1.28

Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 170 °C
Melting Pt: -7 °C
Density: 1.28 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,03171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b28237b7bd4043d9ae23da99b209c1e7e501c05&vid=4aa60dcdb4a23c00058886473425f0353d594a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR