3,6-Dichloropyridazine ≥98.0%

Supplier: TCI America
Warning

D1465-500G D1465-25G
TCD1465-500GEA 253 USD
TCD1465-500G TCD1465-025G
3,6-Dichloropyridazine ≥98.0%
3,6-Dichloropyridazine

CAS Number: 141-30-0
MDL Number: MFCD00006466
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69

Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 69 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1d188683e53a57b94a516d1158ac166c91e11f&vid=ea1d188683e53a57b94a516d1158ac166c91e11f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR