4-(Dimethylamino)pyridine ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

D1450-500G D1450-100G D1450-25G
TCD1450-500GEA 389.27 USD
TCD1450-500G TCD1450-100G TCD1450-25G
4-(Dimethylamino)pyridine ≥99.0% (by titrimetric analysis)
4-(Dimethylamino)pyridine

CAS Number: 1122-58-3
MDL Number: MFCD00006418
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >99.0% (T)
Form: Crystalline Powder
Boiling point (°C): 190
Melting point (°C): 110
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: DMAP, N-(4-Pyridyl)dimethylamine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 190 °C
Melting Pt: 110 °C
Flash Pt: 110 °C
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca7ad9c7c1a19b5b83e5a404f61ac9d5c914575&vid=8ca7ad9c7c1a19b5b83e5a404f61ac9d5c914575&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR