3,5-Dichlorobenzoyl chloride ≥95.0%

Supplier: TCI America

D1404-500G D1404-25G
TCD1404-500GEA 287 USD
TCD1404-500G TCD1404-25G
3,5-Dichlorobenzoyl chloride ≥95.0%
3,5-Dichlorobenzoyl chloride

CAS Number: 2905-62-6
MDL Number: MFCD00009817
Molecular Formula: C7H3Cl3O
Molecular Weight: 209.45
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Flash Point (°C): 137
Freezing point (°C): 24

Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 26 - 28 °C
Density: 1.478 g/cm³ (20 °C)
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00009817
CAS Number: 2905-62-6
EINECS: 220-813-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd62dfdddb0c3f816f9073a00256168916ad46&vid=fabd62dfdddb0c3f816f9073a00256168916ad46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR