Diaminomaleonitrile ≥96.0%
Supplier: TCI America
D1335-500G
D1335-25G
D1335-100G
TCD1335-500GEA
246.42
USD
TCD1335-500G
TCD1335-25G
TCD1335-100G
Diaminomaleonitrile ≥96.0%
Diaminomaleonitrile
CAS Number: 1187-42-4
MDL Number: MFCD00001870
Molecular Formula: C4H4N4
Molecular Weight: 108.10
Purity/Analysis Method: >96.0% (N)
Form: Crystal
Melting point (°C): 180
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 1,2-Diamino-1,2-dicyanoethylene
Formula:
C₄H₄N₄ MW: 108.1 g/mol Boiling Pt: 445 Melting Pt: 180 °C Density: 1.368 g/cm³ (20 °C) |
MDL Number:
MFCD00001870 CAS Number: 1187-42-4 EINECS: 214-697-6 UN: 3439 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...