Didodecyl sulphide ≥96.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Dodecyl sulphide
D1327-25G
D1327-500G
TCD1327-025GEA
62.8
USD
TCD1327-025G
TCD1327-500G
Didodecyl sulphide ≥96.0%
Didodecyl sulphide
CAS Number: 2469-45-6
MDL Number: MFCD00008975
Molecular Formula: C24H50S
Molecular Weight: 370.72
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 41
Flash Point (°C): 110
Other Product Names / Synonyms: Didodecyl Sulfide, Lauryl Sulfide
Formula:
C₂₄H₅₀S MW: 370.73 g/mol Boiling Pt: 465 Melting Pt: 40 Density: 0.844 g/cm³ (30 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00008975 CAS Number: 2469-45-6 EINECS: 219-587-1 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...