5-Decyne ≥98.0%
Supplier: TCI America
D1190-5ML
D1190-25ML
TCD1190-005MLEA
88
USD
TCD1190-005ML
TCD1190-25ML
5-Decyne ≥98.0%
5-Decyne
CAS Number: 1942-46-7
MDL Number: MFCD00027277
Molecular Formula: C10H18
Molecular Weight: 138.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 54
Specific Gravity (20/20): 0.77
Formula:
C₁₀H₁₈ Boiling Pt: 177 °C Density: 0.77 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00027277 CAS Number: 1942-46-7 UN: 3295 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...