4,4-Dimethoxybutanone ≥90.0%

Supplier: TCI America

Synonyms: 4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone

D1157-500ML D1157-25ML D1157-100ML
TCD1157-500MLEA 168 USD
TCD1157-500ML TCD1157-25ML TCD1157-100ML
4,4-Dimethoxybutanone ≥90.0%
4,4-Dimethoxybutanone

CAS Number: 5436-21-5
MDL Number: MFCD00008789
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 58
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Acetoacetaldehyde Dimethyl Acetal

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 170 °C
Melting Pt: –82 °C
Density: 1.00 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3a7e84dbaf0b99cd39c7f48ca00fbb62588416&vid=dc3a7e84dbaf0b99cd39c7f48ca00fbb62588416&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR