2,3-Dimethyl-1,3-butadiene ≥98.0% stabilized

Supplier: TCI America

Synonyms: 2,3-Dimethylbuta-1,3-diene

Stabilization: Stabilised with Stabilised

D1148-10ML D1148-25ML
TCD1148-010MLEA 61.01 USD
TCD1148-010ML TCD1148-25ML
2,3-Dimethyl-1,3-butadiene ≥98.0% stabilized
2,3-Dimethyl-1,3-butadiene

(stabilized with BHT)
CAS Number: 513-81-5
MDL Number: MFCD00008595
Molecular Formula: C6H10
Molecular Weight: 82.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Flash Point (°C): -1
Specific Gravity (20/20): 0.73
Storage Temperature: <0°C

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 68 °C
Melting Pt: -76 °C
Density: 0.73 (20 °C)
Flash Pt: -1 °C
MDL Number: MFCD00008595
CAS Number: 513-81-5
EINECS: 208-172-0
UN: 3295
ADR: 3,II
Merck Index: 13,03265

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR