2,3-Dimethyl-1-butene ≥98.0%
Supplier: TCI America
D1139-25ML
D1139-500ML
TCD1139-025MLEA
93.77
USD
TCD1139-025ML
TCD1139-500ML
2,3-Dimethyl-1-butene ≥98.0%
2,3-Dimethyl-1-butene
CAS Number: 563-78-0
MDL Number: MFCD00008923
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 55
Flash Point (°C): -18
Specific Gravity (20/20): 0.68
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 55 °C Melting Pt: –157 °C Density: 0.68 (20 °C) Flash Pt: –18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008923 CAS Number: 563-78-0 EINECS: 209-262-2 UN: 3295 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...