2,4-Dichloro-3,5-xylenol ≥98.0%
Supplier: TCI America
CAS Number: 133-53-9
MDL Number: MFCD00019981
Molecular Formula: C8H8Cl2O
Molecular Weight: 191.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93
Other Product Names / Synonyms: 2,4-Dichloro-m-xylenol
Formula:
C₈H₈Cl₂O Boiling Pt: 269 °C (1013 hPa) Melting Pt: 93 °C |
MDL Number:
MFCD00019981 CAS Number: 133-53-9 EINECS: 205-109-9 Merck Index: 13,03102 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...