Troclosene sodium ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

D1003-500G D1003-25G
TCD1003-500GEA 43.9 USD
TCD1003-500G TCD1003-25G
Troclosene sodium ≥97.0% (by titrimetric analysis)
Troclosene sodium

CAS Number: 2893-78-9
MDL Number: MFCD00006036
Molecular Formula: C3HCl2N3O3
Molecular Weight: 219.94
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Dichloroisocyanuric Acid Sodium Salt

Formula: C₃Cl₂N₃NaO₃
Melting Pt: 225 °C
Density: 0.96 g/cm³ (20 °C)
MDL Number: MFCD00006036
CAS Number: 2893-78-9
EINECS: 220-767-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a433fd80d9274162ded82e53eb624c5c22c751&vid=a7a433fd80d9274162ded82e53eb624c5c22c751&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR