1-Dodecene ≥95.0%

Supplier: TCI America

Synonyms: Dodec-1-ene

D0974-25ML D0974-500ML
TCD0974-025MLEA 22.39 USD
TCD0974-025ML TCD0974-500ML
1-Dodecene ≥95.0%
1-Dodecene

CAS Number: 112-41-4
MDL Number: MFCD00008961
Molecular Formula: C12H24
Molecular Weight: 168.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 213
Melting point (°C): -35
Flash Point (°C): 76
Specific Gravity (20/20): 0.76

Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 213 °C
Melting Pt: –35 °C
Density: 0.76 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23696b80fb70918d96eda909f3c7ba0422012b05&vid=40edcd1a7d449ad5e51e2a4af1709e9d0d00432e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR