Lauroyl chloride ≥98.0%

Supplier: TCI America

Synonyms: Dodecanoyl chloride

D0972-500ML D0972-25ML
TCD0972-500MLEA 81.53 USD
TCD0972-500ML TCD0972-25ML
Lauroyl chloride ≥98.0%
Lauroyl chloride

CAS Number: 112-16-3
MDL Number: MFCD00000740
Molecular Formula: C12H23ClO
Molecular Weight: 218.77
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 132
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Dodecanoyl Chloride

Formula: C₁₂H₂₃ClO
MW: 218.77 g/mol
Boiling Pt: 145 °C
Melting Pt: -12 °C
Density: 0.92 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00000740
CAS Number: 112-16-3
EINECS: 203-941-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed740ddd73e993eee1af90520f6b9d4782579bc&vid=eed740ddd73e993eee1af90520f6b9d4782579bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR