Divinylbenzene (mixture of isomers) 50.0% (contains Ethylvinylbenzene, Diethylbenzene) stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

D0958-500ML D0958-25ML
TCD0958-500MLEA 76.99 USD
TCD0958-500ML TCD0958-025ML
Divinylbenzene (mixture of isomers) 50.0% (contains Ethylvinylbenzene, Diethylbenzene) stabilized
Divinylbenzene (mixture of isomers)

(stabilized with TBC)
CAS Number: 1321-74-0
MDL Number: MFCD01778656
Molecular Formula: C10H10
Molecular Weight: 130.19
Purity/Analysis Method: >50.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 68
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Formula: C₁₀H₁₀
Boiling Pt: 195 °C
Density: 0.91 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5

Order Now

Specification Test Results

Purity/Analysis Method >50.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR