Diphenylmethane ≥99.0%

Supplier: TCI America
Warning

D0896-500G D0896-25G
TCD0896-500GEA 49.84 USD
TCD0896-500G TCD0896-025G
Diphenylmethane ≥99.0%
Diphenylmethane

CAS Number: 101-81-5
MDL Number: MFCD00004781
Molecular Formula: C13H12
Molecular Weight: 168.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 130
Freezing point (°C): 25


Other Product Names / Synonyms: Benzylphenyl

Formula: C₁₃H₁₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 22…25 °C
Density: 0.9985 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004781
CAS Number: 101-81-5
EINECS: 202-978-6
UN: 3077
ADR: 9,III
Merck Index: 13,03360
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81a22d8738bc5d7fe65aaa4d374e645de08aa538&vid=81a22d8738bc5d7fe65aaa4d374e645de08aa538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR