2,4-Dinitrotoluene ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1,3-Dinitro-4-methylbenzene

D0856-500G D0856-25G
TCD0856-500GEA 110 USD
TCD0856-500G TCD0856-25G
2,4-Dinitrotoluene ≥99.0%
2,4-Dinitrotoluene

CAS Number: 121-14-2
MDL Number: MFCD00007172
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 71
Flash Point (°C): 207

Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Boiling Pt: 300 °C
Melting Pt: 71 °C
Density: 1.379 g/cm³ (20 °C)
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00007172
CAS Number: 121-14-2
EINECS: 204-450-0
UN: 3454
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74915c0e62624f0e3f8cc61a9af9b0b4978711a0&vid=181c0bc918f4cbc525517df6453b5e15160afd89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR