1,4-Dimethylpiperazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: sym-Dimethylpiperazine

D0787-500ML D0787-25ML
TCD0787-500MLEA 145 USD
TCD0787-500ML TCD0787-25ML
1,4-Dimethylpiperazine ≥98.0% (by GC, titration analysis)
1,4-Dimethylpiperazine

CAS Number: 106-58-1
MDL Number: MFCD00006156
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 133
Melting point (°C): -1
Flash Point (°C): 25
Specific Gravity (20/20): 0.85

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 133 °C
Melting Pt: -1 °C
Density: 0.85 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00006156
CAS Number: 106-58-1
EINECS: 203-412-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7167d491356de0f3dbfa61eea92e9885dd026001&vid=7167d491356de0f3dbfa61eea92e9885dd026001&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR