N,N-Dimethyl-p-phenylenediammonium sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: N,N-Dimethyl-1,4-phenylenediammonium sulphate, N,N-Dimethylbenzene-1,4-diammonium sulphate

D0782-500G D0782-25G
TCD0782-500GEA 1034 USD
TCD0782-500G TCD0782-25G
N,N-Dimethyl-p-phenylenediammonium sulfate ≥98.0% (by titrimetric analysis)
N,N-Dimethyl-p-phenylenediammoniumsulphate

CAS Number: 536-47-0
MDL Number: MFCD00012992
Molecular Formula: C8H12N2
Molecular Weight: 234.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 205
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Aminodimethylaniline Sulfate

Formula: C₈H₁₄N₂O₄S
MW: 234.28 g/mol
Melting Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012992
CAS Number: 536-47-0
EINECS: 208-636-2
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59ac8efbbdc97c4f99842d96ad20bfb0fe7c84ba&vid=59ac8efbbdc97c4f99842d96ad20bfb0fe7c84ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR