2,3-Xylenol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2,3-Dimethylphenol, 3-Hydroxy-2-xylene, 3-Hydroxy-o-xylene
D0773-500G
D0773-25G
TCD0773-500GEA
324
USD
TCD0773-500G
TCD0773-25G
2,3-Xylenol ≥98.0%
2,3-Xylenol
CAS Number: 526-75-0
MDL Number: MFCD00002227
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 218
Melting point (°C): 75
Flash Point (°C): 95
Other Product Names / Synonyms: 2,3-Xylenol
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 218 °C Melting Pt: 75 °C Density: 0.965 g/cm³ (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00002227 CAS Number: 526-75-0 EINECS: 208-395-3 UN: 2261 ADR: 6.1,II Merck Index: 12,10215 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...