2,4-Dimethyl-3-pentanone ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 565-80-0
MDL Number: MFCD00008918
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Melting point (°C): -80
Flash Point (°C): 18
Specific Gravity (20/20): 0.80
Other Product Names / Synonyms: Diisopropyl Ketone
Formula:
C₇H₁₄O Boiling Pt: 125 °C Melting Pt: –80 °C Density: 0.80 (20 °C) Flash Pt: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008918 CAS Number: 565-80-0 EINECS: 209-294-7 UN: 1224 ADR: 3,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...