N,N-Dimethylethylendiamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

D0719-500ML D0719-25ML
TCD0719-500MLEA 263.39 USD
TCD0719-500ML TCD0719-25ML
N,N-Dimethylethylendiamine ≥98.0% (by GC, titration analysis)
N,N-Dimethylethylendiamine

CAS Number: 108-00-9
MDL Number: MFCD00008175
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 18
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: 2-Dimethylaminoethylamine

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: 107 °C
Melting Pt: –70 °C
Density: 0.81 (20 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df54a1e81bb6d39f938c29d5703ffc56188721db&vid=df54a1e81bb6d39f938c29d5703ffc56188721db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR