Benzyl(dimethyl)amine ≥98.0%

Supplier: TCI America
Danger

Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

D0688-500ML D0688-25ML
TCD0688-500MLEA 50.01 USD
TCD0688-500ML TCD0688-25ML
Benzyl(dimethyl)amine ≥98.0%
Benzyl(dimethyl)amine

CAS Number: 103-83-3
MDL Number: MFCD00008329
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Melting point (°C): -75
Flash Point (°C): 57
Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: N-Benzyldimethylamine, DMBA, BDMA

Formula: C₆H₅CH₂N(CH₃)₂
MW: 135.21 g/mol
Boiling Pt: 180 °C
Melting Pt: –75 °C
Density: 0.90 (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008329
CAS Number: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07455d8ea132474ef40eba608506f8b1906d4146&vid=07455d8ea132474ef40eba608506f8b1906d4146&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR